missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-oxazoline, 98%

Product Code. 11480543
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480543 5 g 5g
11490543 25 g 25g
11400553 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12670.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 30093-99-3 | C5H9NO | 99.13 g/mol

4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30093-99-3
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00014507
InChI Key KOAMXHRRVFDWRQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl
PubChem CID 121630
IUPAC Name 4,4-dimethyl-5H-1,3-oxazole
SMILES CC1(C)COC=N1

Specifications

Density 0.94
Boiling Point 99°C to 100°C
Flash Point 1°C (33°F)
Refractive Index 1.419
Quantity 5 g
UN Number UN1993
Beilstein 1209606
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 99.13
Percent Purity 98%
Chemical Name or Material 4,4-Dimethyl-2-oxazoline
Show More Show Less
compliance-icons
Product Identifier
  • 4,4-Dimethyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: 4,4-Dimethyl-2-oxazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.