missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-oxazoline, 98%

Product Code. 10313872 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10313872 10 g Glass bottle 10g
11976821 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10313872 Supplier Thermo Scientific Acros Supplier No. 355270100
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 30093-99-3 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30093-99-3
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00014507
InChI Key KOAMXHRRVFDWRQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-yl
PubChem CID 121630
SMILES CC1(C)COC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9400g/mL
Boiling Point 99.0°C to 100.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4240
Quantity 10 g
Specific Gravity 0.94
IUPAC Name 4,4-dimethyl-4,5-dihydro-1,3-oxazole
Formula Weight 99.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4, 4-Dimethyl-2-oxazoline
Show More Show Less
compliance-icons
Product Identifier
  • 4,4-Dimethyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.