missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester, 98+%

Product Code. 15441146
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441146 100 mg 100mg
15431146 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441146 Supplier Thermo Scientific Alfa Aesar Supplier No. H54402.MC
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 146998-31-4 | C19H18N4O4 | 366.377 g/mol

4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester is a useful reagent (similar to MANCYL-SE) for labeling proteins or peptides through their amino-groups by forming stable peptide bonds. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles for fluorescent studies of proteins. Its fluorescent properties (has a characteristic broad and intense visible absorption but has no fluorescence) make it an ideal long wavelength quencher and it has been utilized as an acceptor chromophore in FRET studies. It is also used for N-terminal modification of peptides in automated synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester is a useful reagent (similar to MANCYL-SE) for labeling proteins or peptides through their amino-groups by forming stable peptide bonds. The succinimidyl ester is reactive with terminal amines or lysines of peptides and other nucleophiles for fluorescent studies of proteins. Its fluorescent properties (has a characteristic broad and intense visible absorption but has no fluorescence) make it an ideal long wavelength quencher and it has been utilized as an acceptor chromophore in FRET studies. It is also used for N-terminal modification of peptides in automated synthesis.

Solubility
Soluble in DMSO.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146998-31-4
Molecular Formula C19H18N4O4
Molecular Weight (g/mol) 366.377
MDL Number MFCD00467415
InChI Key IBOVDNBDQHYNJI-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, dabcyl se dye, dabcyl acid, se, 4-4-dimethylamino phenyl azo benzoic acid, succinimidyl ester, 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl-nhs, e-2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, 2,5-pyrrolidinedione, 1-4-4-dimethylamino phenyl azo benzoyl oxy, dabcyl-se; 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl se, solid
PubChem CID 4206604
ChEBI CHEBI:52005
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)ON3C(=O)CCC3=O

Spezifikation

Quantity 100 mg
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO.
Formula Weight 366.37
Percent Purity ≥98%
Chemical Name or Material 4-[4-(Dimethylamino)phenylazo]benzoic acid N-succinimidyl ester

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.