missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexan-1-ol, 95%, Thermo Scientific™

Product Code. 10600245
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10600245 1 g Amber Glass Bottle 1g
10547473 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10600245 Supplier Thermo Scientific Maybridge Supplier No. BTBG00026DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 932-01-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00101954
InChI Key VUQOIZPFYIVUKD-UHFFFAOYSA-N
PubChem CID 136735
IUPAC Name 4,4-dimethylcyclohexan-1-ol
SMILES CC1(C)CCC(O)CC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4,4-dimethylcyclohexanol, cyclohexanol, 4,4-dimethyl, 4,4-dimethyl-cyclohexanol, cyclohexanol,4,4-dimethyl, 4,4-dimethyl-1-cyclohexanol, 4,4-dimethyl cyclohexan-1-ol
Formula Weight 128.21
Percent Purity 95%
Chemical Name or Material 4, 4-Dimethylcyclohexan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.