missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dithiodipyridine, 98%

Product Code. 10607332 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10607332 25 g Glass bottle 25g
10307390 1 g Glass bottle 1g
10478530 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607332 Supplier Thermo Scientific Acros Supplier No. 162240250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2645-22-9 | C10H8N2S2 | 220.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2645-22-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.32
MDL Number MFCD00006423
InChI Key UHBAPGWWRFVTFS-UHFFFAOYSA-N
Synonym 4,4'-dithiodipyridine, 4,4'-dipyridyl disulfide, 1,2-di pyridin-4-yl disulfane, aldrithiol-4, pyridine, 4,4'-dithiobis, 4,4'-dithiopyridine, di 4-pyridyl disulfide, aldrithiol 4, di 4-pyridinyl disulfide, 4,4'-dipyridine disulfide
PubChem CID 75846
ChEBI CHEBI:41814
IUPAC Name 4-(pyridin-4-yldisulfanyl)pyridine
SMILES C1=CN=CC=C1SSC2=CC=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 79°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)S-S(C5H4N)
Quantity 25 g
Beilstein 21, II, 35
Formula Weight 220.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Dithiodipyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dithiodipyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.