missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)imidazole, 97%

Product Code. 15414107
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414107 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414107 Supplier Thermo Scientific Alfa Aesar Supplier No. H31503.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65020-70-4 | C9H7FN2 | 162.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 65020-70-4
Melting Point 127°C to 131°C
Molecular Formula C9H7FN2
MDL Number MFCD00297041
Quantity 5 g
Synonym 4-4-fluorophenyl-1h-imidazole, 5-4-fluorophenyl-1h-imidazole, 1h-imidazole,5-4-fluorophenyl, 4-4-fluoro-phenyl-1h-imidazole, regid_for_cid_657876, 5-4-fluorophenyl-3h-imidazole, 4-4-fluorophenyl-1h-imidazole, 9, 1h-imidazole, 5-4-fluorophenyl, 3sq
InChI Key QYDAFWKRBZYBOB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=CN=CN2)F
IUPAC Name 5-(4-fluorophenyl)-1H-imidazole
Molecular Weight (g/mol) 162.167
PubChem CID 657876
Formula Weight 162.17
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenyl)imidazole
Show More Show Less

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.