missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Isocyanatophenoxy)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 10723854
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723854 1 g Amber Glass Bottle 1g
10003123 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723854 Supplier Thermo Scientific Maybridge Supplier No. CC52406DA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 892501-94-9
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD09702366
InChI Key ZGNHUEWFJQLPDN-UHFFFAOYSA-N
Synonym 4-4-isocyanatophenoxy oxane, 4-4-isocyanatophenoxy tetrahydropyran, 4-4-isocyanatophenoxy tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-4-isocyanatophenoxy, 4-tetrahydro-2h-pyran-4-yl oxy phenyl isocyanate, 1-isocyanato-4-tetrahydro-2h-pyran-4-yl oxy benzene, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenisocyanate
PubChem CID 24229521
IUPAC Name 4-(4-isocyanatophenoxy)oxane
SMILES C1COCCC1OC2=CC=C(C=C2)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material 4-(4-Isocyanatophenoxy)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.