missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™

Product Code. 11905691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11905691 100 mL AcroSeal™ Glass Bottle 100mL
11995681 1 L AcroSeal™ Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905691 Supplier Thermo Scientific Acros Supplier No. 431791000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5124-30-1 | C15H22N2O2 | 262.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5124-30-1
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD00015405
InChI Key KORSJDCBLAPZEQ-UHFFFAOYSA-N
Synonym hydrogenated mdi, hylene w, bis 4-isocyanatocyclohexyl methane, 4,4'-diisocyanatodicyclohexylmethane, methylene bis 4-cyclohexylisocyanate, dicyclohexylmethane 4,4'-diisocyanate, dicyclohexylmethane-4,4'-diisocyanate, methylene bis-4-cyclohexylisocyanate, nacconate h 12, cyclohexane, 1,1'-methylenebis 4-isocyanato
PubChem CID 21202
ChEBI CHEBI:53216
IUPAC Name 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
SMILES O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 19.0°C to 23.0°C
Color Colorless
Density 1.0660g/mL
Boiling Point 112°C to 114°C (1010.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 1.066
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone
Formula Weight 262.35
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4, 4'-Methylenebis(cyclohexyl isocyanate)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(cyclohexyl isocyanate)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.