missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(phenyl Isocyanate), TRC

Product Code. 30472433
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472433 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472433 Supplier Toronto Research Chemicals Supplier No. TRCM2273801G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylmethan-4,4'-diisocyanate
CAS 101-68-8
Molecular Formula C15 H10 N2 O2
Quantity 1 g
Synonym Isocyanic acid, methylenedi-p-phenylene ester (6CI,8CI), 1,1'-Methylenebis[4-isocyanatobenzene], 4,4'-Bis(isocyanatophenyl)methane, 4,4'-Diisocyanatodiphenylmethane, 4,4'-Diphenylmethane diisocyanate, 4,4'-MDI, 4,4'-Methylenebis(isocyanatobenzene), 4,4'-Methylenebis(phenyl isocyanate), 4,4'-Methylenebis(phenylisocyante), 4,4'-Methylenedi(phenyl isocyanate), 4,4'-Methylenedi-p-phenylene diisocyanate, 4,4'-Methylenediphenyl diisocyanate, 4,4'-Methylenediphenylene isocyanate, Bis(1,4-isocyanatophenyl)methane, Bis(4-isocyanatophenyl)methane, Bis(isocyanatophenyl)methane, Bis(p-isocyanatophenyl)methane, Di(4-isocyanatophenyl)methane, Diisocyanatodiphenylmethane, Diphenylmethane 4,4'-diisocyanate, Diphenylmethane diisocyanate, Diphenylmethyl diisocyanate, Diphenylmethylene 4,4'-diisocyanate, Diphenylmethylene diisocyanate, Isocyanic acid, diphenylmethylene ester, MDI, Methylbis(4-phenylisocyanate), Methyldi-p-phenylene isocyanate, Methylene diisocyanate, Methylene diphenyl diisocyanate, Methylenebis(4-isocyanatobenzene), Methylenebis(4-phenyl isocyanate), Methylenebis(4-phenylene isocyanate), Methylenebis(p-phenyl isocyanate), Methylenebis(p-phenylene isocyanate), Methylenebis(phenyl isocyanate), Methylenebis-p-phenylene diisocyanate, Methylenebisphenylene diisocyanate, Methylenedi-p-phenylene diisocyanate, Methylenedi-p-phenylene isocyanate, Methylenediphenylisocyanate, NSC 9596, p,p'-Diphenylmethane diisocyanate, p,p'-Methylenebis(phenyl isocyanate), p,p'-Methylenediphenyl diisocyanate, Diphenylmethan-4,4'-diisocyanate
InChI Formula InChI=1S/C15H10N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h1-8H,9H2
SMILES O=C=Nc1ccc(Cc2ccc(cc2)N=C=O)cc1
IUPAC Name 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
Molecular Weight (g/mol) 250.25
Formula Weight 250.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.