missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Methylenedianiline-2,2',6,6',N,N,N',N'-d8, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4,4'-Diaminodiphenylmethane-N,N,N',N',3,3',5,5'-D8 |
| CAS | 1219795-26-2 |
| Molecular Formula | C13 D8 H6 N2 |
| Quantity | 0.1 g |
| Synonym | Benzen-2,6-d2-amine-d2, 4,4'-methylenebis- (ACI), 4,4'-Methylenebis[benzen-2,6-d2-amine-d2] (ACI), 4,4'-Methylenedianiline-2,2',6,6',N,N,N',N'-d8, Bis-(4-aminophenyl-N,N,3,5-d4)methane, 4,4'-Diaminobiphenylmethane-d8, 4,4'-Diaminodiphenylmethane-N,N,N',N',3,3',5,5'-D8 |
| InChI Formula | InChI=1 S/C13H14N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8 H,9,14-15H2/i5D,6 D,7 D,8 D/hD4 |
| SMILES | [2 H]N([2 H])c1c([2 H])cc(Cc2cc([2 H])c(N([2 H])[2 H])c([2 H])c2)cc1[2 H] |
| IUPAC Name | N,N,2,6-tetradeuterio-4-[[3,5-dideuterio-4-(dideuterioamino)phenyl]methyl]aniline |
| Molecular Weight (g/mol) | 206.313 |
| Formula Weight | 206.1659 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction