missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Methylpiperazino)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Quantity | 5 g |
| Formula Weight | 204.27 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(4-Methylpiperazino)benzaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction