missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazinomethyl)benzoic Acid, Dihydrochloride, TRC

Product Code. 30472167
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472167 1 g 1g
30479760 2.5 g 2.5g
30467293 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472167 Supplier Toronto Research Chemicals Supplier No. TRCM3259211G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106261-49-8
Molecular Formula C13 H18 N2 O2 . 2 Cl H
Molecular Weight (g/mol) 307.22
Synonym 4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid Hydrochloride (1:2), Benzoic acid, 4-[(4-methyl-1-piperazinyl)methyl]-, dihydrochloride (9CI), 4-(4-Methylpiperazinomethyl)benzoic acid dihydrochloride, 4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride, 4-[(4-Methylpiperazinyl)methyl]benzoic acid dihydrochloride
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid;dihydrochloride
SMILES Cl.Cl.CN1CCN(Cc2ccc(cc2)C(=O)O)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H18N2O2.2ClH/c1-14-6-8-15(9-7-14)10-11-2-4-12(5-3-11)13(16)17;;/h2-5H,6-10H2,1H3,(H,16,17);2*1H
Formula Weight 306.0902
Chemical Name or Material 4-(4-Methylpiperazinomethyl)benzoic Acid, Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.