missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)aniline, 98+%

Product Code. 11468780
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468780 5 g 5g
15443217 1 g 1g
11478780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468780 Supplier Thermo Scientific Alfa Aesar Supplier No. B20292.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2524-67-6 | C10H14N2O | 178.235 g/mol

4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

Solubility
Soluble in chloroform, ethyl acetate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-67-6
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00006169
InChI Key PHNDZBFLOPIMSM-UHFFFAOYSA-N
Synonym 4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
PubChem CID 75655
IUPAC Name 4-morpholin-4-ylaniline
SMILES C1COCCN1C2=CC=C(C=C2)N

Specifications

Melting Point 129°C to 133°C
Quantity 5 g
UN Number UN2811
Beilstein 150957
Solubility Information Soluble in chloroform,ethyl acetate.
Formula Weight 178.24
Percent Purity ≥98%
Chemical Name or Material 4-(4-Morpholinyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.