missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrobenzyl)pyridine, 98%

Product Code. p-3758082 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573481 5 g Glass bottle 5g
10376530 10 g Glass bottle 10g
10454575 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10573481 Supplier Thermo Scientific Acros Supplier No. 128550050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 1083-48-3 | C12H10N2O2 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1083-48-3
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00006445
InChI Key MNHKUCBXXMFQDM-UHFFFAOYSA-N
Synonym 4-4-nitrobenzyl pyridine, 4-4-nitrophenyl methyl pyridine, pyridine, 4-4-nitrophenyl methyl, pyridine, 4-p-nitrobenzyl, 1-p-nitrobenzyl pyridine, 4-p-nitrobenzyl pyridine, 4 4-nitrobenzyl primide, unii-0z6gs37n7h, gamma-p-nitrobenzyl pyridine
PubChem CID 14129
IUPAC Name 4-[(4-nitrophenyl)methyl]pyridine
SMILES [O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1

Spezifikation

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)CH2C6H4NO2
Quantity 5 g
Beilstein 20, II, 272
Solubility Information Solubility in water: 1,04g/L
Formula Weight 214.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-(4-Nitrobenzyl)pyridine
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 4-(4-Nitrobenzyl)pyridine
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.