missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenyl)-3-morpholinone, TRC

Product Code. 30473063
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473063 100 mg -
30476838 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473063 Supplier Toronto Research Chemicals Supplier No. TRCN504425100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446292-04-2
Molecular Formula C10 H10 N2 O4
Molecular Weight (g/mol) 222.2
Synonym 4-(4-Nitrophenyl)-3-morpholinone, 4-(3-Oxo-4-morpholinyl)nitrobenzene, 4-(4-Nitrophenyl)-3-oxomorpholine, 4-(4-Nitrophenyl)morpholin-3-one
IUPAC Name 4-(4-nitrophenyl)morpholin-3-one
SMILES [O-][N+](=O)c1ccc(cc1)N2CCOCC2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H10N2O4/c13-10-7-16-6-5-11(10)8-1-3-9(4-2-8)12(14)15/h1-4H,5-7H2
Formula Weight 222.0641
Chemical Name or Material 4-(4-Nitrophenyl)-3-morpholinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.