missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10344702
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
Product Code. Quantity Packaging unitSize
10344702 250 mg Amber Glass Bottle 250mg
10059191 1 g Amber Glass Bottle 1g
10615145 10 g Amber Glass Bottle 10g
3 options
This item is not returnable. View return policy

Product Code. 10344702

Brand: Thermo Scientific Maybridge BTB01214CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 27692-74-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00053060
InChI Key WZXYYQHVDJMIFF-UHFFFAOYSA-N
Synonym 4-4-pyridylmethyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, 4-4-pyridinylmethyl aniline, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, enamine_005904
PubChem CID 307158
IUPAC Name 4-(pyridin-4-ylmethyl)aniline
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 184.24
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-(4-Pyridylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.