missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(Trifluoromethoxy)phenoxy]piperidine, TRC

Product Code. 30453715
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453715 Supplier Toronto Research Chemicals Supplier No. TRCT7908901G
 Restricted Item
Estimated Shipment: 09-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
CAS 287952-67-4
Molecular Formula C12H14F3NO2
Quantity 1 g
InChI Formula InChI=1S/C12H14F3NO2/c13-12(14,15)18-11-3-1-9(2-4-11)17-10-5-7-16-8-6-10/h1-4,10,16H,5-8H2
SMILES FC(F)(F)Oc1ccc(OC2CCNCC2)cc1
IUPAC Name 4-[4-(trifluoromethoxy)phenoxy]piperidine
Molecular Weight (g/mol) 261.24
Formula Weight 261.0977
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.