missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]morpholine, Thermo Scientific™

Product Code. 10611875
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10611875 1 g Amber Glass Bottle 1g
11370577 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10611875 Supplier Thermo Scientific Maybridge Supplier No. CC17539DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 485799-04-0
Molecular Formula C15H23BN2O3
Molecular Weight (g/mol) 290.17
MDL Number MFCD04039874
InChI Key ZGDLVKWIZHHWIR-UHFFFAOYSA-N
Synonym 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholin-4-yl pyridine-3-boronic acid pinacol ester, 6-morpholinopyridin-3-ylboronic acid pinacol ester, 2-morpholinopyridine-5-boronic acid, pinacol ester, 4-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-morpholine, 4-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl morpholine, 2-4-morpholino pyridine-5-boronic acid pinacol ester, 4-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl morpholine, 6-morpholinopyridine-3-boronic acid pinacol ester
PubChem CID 2795363
IUPAC Name 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 290.17
Percent Purity 97%
Chemical Name or Material 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.