missing translation for 'onlineSavingsMsg'
Learn More

4,5,6-Triaminopyrimidine sulfate hydrate, 98+% (dry wt.), water <8%

Product Code. 11419317
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419317 5 g 5g
11409317 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419317 Supplier Thermo Scientific Alfa Aesar Supplier No. B21369.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 207742-76-5 | C4H9N5O4S | 223.21 g/mol

4,5,6-Triaminopyrimidine sulfate hydrate is used to prepare 2,5-anhydro-3-deoxy-N-(4',6'-diaminopyrimidin-5'yl)-D-ribo-hexonamide by reaction with 2,5-anhydro-4-O-benzyl-3-deoxy-D-ribo-hexonic acid. It is also used to prepare 4,5,6-triaminopyrimidine sulfite. Further, it acts as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5,6-Triaminopyrimidine sulfate hydrate is used to prepare 2,5-anhydro-3-deoxy-N-(4′,6′-diaminopyrimidin-5′yl)-D-ribo-hexonamide by reaction with 2,5-anhydro-4-O-benzyl-3-deoxy-D-ribo-hexonic acid. It is also used to prepare 4,5,6-triaminopyrimidine sulfite. Further, it acts as an intermediate in synthetic chemistry.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207742-76-5
Molecular Formula C4H9N5O4S
Molecular Weight (g/mol) 223.21
MDL Number MFCD00012789
InChI Key RKJICTKHLYLPLY-UHFFFAOYSA-N
Synonym 4,5,6-triaminopyrimidine sulfate hydrate, 4,5,6-triaminepyrimidine sulfate hydrate, pyrimidine, 4,5,6-triamino-, sulfate, hydrate, pyrimidine-4,5,6-triamine sulfate hydrate, pyrimidine-4,5,6-triamine sulfuric acid hydrate, pyrimidine-4,5,6-triamine; sulfuric acid; hydrate, zlchem 873, acmc-209nvb, c4h7n5.h2o.h2so4, 4,5,6-triaminopyrimidine sulfate, hydrate
PubChem CID 201714
IUPAC Name pyrimidine-4,5,6-triamine;sulfuric acid;hydrate
SMILES OS(O)(=O)=O.NC1=NC=NC(N)=C1N

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 223.22 (Anhydrous)
Percent Purity ≥98%
Assay (dry wt.), water <8%
Chemical Name or Material 4,5,6-Triaminopyrimidine sulfate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.