missing translation for 'onlineSavingsMsg'
Learn More

4,5-Diaminopyrimidine, 97%

Product Code. 10366354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10366354 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366354 Supplier Thermo Scientific Acros Supplier No. 112400010
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13754-19-3 | C4H6N4 | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13754-19-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 200°C
Color Colorless
Boiling Point 229°C (32.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H6N4
MDL Number MFCD00006113
Quantity 1 g
Beilstein 24, 324
Synonym 4,5-diaminopyrimidine, 4,5-pyrimidinediamine, pyrimidine, 4,5-diamino, 4,5-pyrimidinediamine 9ci, pubchem7057, pyrimidine,5-diamino, 4,5 diamino pyrimidine, 4,5-diamino-pyrimidine, acmc-1cfbq, 4,5-diaminopyrimidiine
InChI Key PPAULTVPKLVLII-UHFFFAOYSA-N
SMILES NC1=CN=CN=C1N
IUPAC Name pyrimidine-4,5-diamine
Molecular Weight (g/mol) 110.12
PubChem CID 83703
Formula Weight 110.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 5-Diaminopyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Diaminopyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.