missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™

Product Code. 10110962
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10110962 1 g Amber Glass Bottle 1g
10714763 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10110962 Supplier Thermo Scientific Maybridge Supplier No. BTB07379DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14305-08-9
Molecular Formula C10H6Br2N2O
Molecular Weight (g/mol) 329.979
MDL Number MFCD00052907
InChI Key NQJXEMRMTOSSBQ-UHFFFAOYSA-N
Synonym 4,5-dibromo-2-phenylpyridazin-3 2h-one, 4,5-dibromo-2-phenyl-2,3-dihydropyridazin-3-one, 4,5-dibromo-2-phenyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dibromo-2-phenyl, 4,5-dibromo-2-phenyl-3-pyridazinone, 4,5-dibromo-2-phenyl-pyridazin-3-one, 2-phenyl-4,5-dibromo-3 2h-pyridazinone, 3 2h-pyridazinone,4,5-dibromo-2-phenyl, 4,5-bis bromanyl-2-phenyl-pyridazin-3-one, 4,5-dibromo-2-phenyl-2-hydropyridazin-3-one
PubChem CID 203396
IUPAC Name 4,5-dibromo-2-phenylpyridazin-3-one
SMILES C1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Br)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 329.98
Percent Purity 97%
Chemical Name or Material 4, 5-Dibromo-2-phenyl-2, 3-dihydropyridazin-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.