missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromothiophene-2-sulfonyl chloride, 97%

Product Code. 11495373
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495373 5 g 5g
11485373 1 g 1g
11405383 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495373 Supplier Thermo Scientific Alfa Aesar Supplier No. A13384.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81606-31-7 | C4HBr2ClO2S2 | 340.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81606-31-7
Molecular Formula C4HBr2ClO2S2
Molecular Weight (g/mol) 340.43
MDL Number MFCD00041439
InChI Key WJYGHWXWQSCONR-UHFFFAOYSA-N
Synonym 4,5-dibromothiophene-2-sulphonyl chloride, 2,3-dibromothiophene-5-sulfonyl chloride, 4,5-dibromo-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 4,5-dibromo, 4,5-dibromothiophene-2-sulfonic acid chloride, acmc-20ams9, buttpark 9350-57, 4,5-dibromo 2-thienyl chlorosulfone, 2,3-dibromothiophene-5-sulfonylchloride, 4,5-dibromothiophene-2-sulfonylchloride
PubChem CID 522755
IUPAC Name 4,5-dibromothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC(Br)=C(Br)S1

Specifications

Melting Point 75°C to 79°C
Quantity 5 g
UN Number UN3261
Beilstein 162960
Sensitivity Moisture sensitive
Formula Weight 340.45
Percent Purity 97%
Chemical Name or Material 4,5-Dibromothiophene-2-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.