missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dicaffeoylquinic acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
4,5-Dicaffeoylquinic acid reduces islet cell apoptosis and improves pancreatic function in type 2 diabetic mice, and has obvious inhibitory activities against yeast α-glucosidase. 4,5-Dicaffeoylquinic acid inhibits prostate cancer cells through cell cycle arrest. 4,5-Dicaffeoylquinic acid also has anti-apoptotic, anti-injury and anti-hepatitis B virus effects.
Specifications
Specifications
| Chemical Name or Material | 4,5-Dicaffeoylquinic acid |
| CAS | 57378-72-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H24O12 |
| Quantity | 5 mg |
| Synonym | Isochlorogenic acid C |
| Solubility Information | DMSO : 50 mg/mL (96.81 mM; Need ultrasonic) |
| SMILES | O=C([C@]1(O)C[C@@H](OC(/C=C/C2=CC=C(O)C(O)=C2)=O)[C@@H](OC(/C=C/C3=CC=C(O)C(O)=C3)=O)[C@H](O)C1)O |
| Molecular Weight (g/mol) | 516.45 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction