missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-(4-methylphenyl)-2,3-dihydropyridazin-3-one, 97%, Thermo Scientific™

Product Code. 10487602
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10487602 10 g 10g
10374222 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10487602 Supplier Thermo Scientific Maybridge Supplier No. BTB07813EA
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 33098-21-4
Molecular Formula C11H8Cl2N2O
Molecular Weight (g/mol) 255.098
MDL Number MFCD00135290
InChI Key OWPCMQKRJFNLHF-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3-one, 4,5-dichloro-2-4-methylphenyl pyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-4-methylphenyl, 4,5-dichloro-2-p-tolyl-2h-pyridazin-3-one, 4,5-dichloro-2-4-methylphenyl-2-hydropyridazin-3-one, 4,5-dichloro-2-p-tolyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-3 2h-pyridazinone, 4,5-dichloro-2-4-methylphenyl pyridazin-3 2h-one, 4,5-dichloro-2-4-methylphenyl-2,3-dihydropyridazin-3one
PubChem CID 2774757
IUPAC Name 4,5-dichloro-2-(4-methylphenyl)pyridazin-3-one
SMILES CC1=CC=C(C=C1)N2C(=O)C(=C(C=N2)Cl)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 255.1
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloro-2-(4-methylphenyl)-2, 3-dihydropyridazin-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.