missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroimidazole, 98%

Product Code. 11362436
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362436 5 g 5g
11372436 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362436 Supplier Thermo Scientific Alfa Aesar Supplier No. L06262.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15965-30-7 | C3H2Cl2N2 | 136.96 g/mol

4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15965-30-7
Molecular Formula C3H2Cl2N2
Molecular Weight (g/mol) 136.96
MDL Number MFCD00005195
InChI Key QAJJXHRQPLATMK-UHFFFAOYSA-N
Synonym 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa
PubChem CID 140018
IUPAC Name 4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=CN1

Specifications

Melting Point 179°C to 182°C
Quantity 5 g
Beilstein 606155
Solubility Information Insoluble in water.
Formula Weight 136.97
Percent Purity 98%
Chemical Name or Material 4,5-Dichloroimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.