missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloroisothiazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10069231
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
10069231 250 mg 250mg
10713233 1 g 1g
10701997 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 10069231

Brand: Thermo Scientific Maybridge BTB12548CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 131947-13-2
Molecular Formula C4HCl2NO2S
Molecular Weight (g/mol) 198.017
MDL Number MFCD00186468
InChI Key ZFEHQZVNKOESSZ-UHFFFAOYSA-N
PubChem CID 1244565
IUPAC Name 4,5-dichloro-1,2-thiazole-3-carboxylic acid
SMILES C1(=C(SN=C1C(=O)O)Cl)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 4,5-dichloroisothiazole-3-carboxylic acid, 4,5-dichloro-isothiazole-3-carboxylic acid, 3-isothiazolecarboxylicacid, 4,5-dichloro, dichloro-1,2-thiazole-3-carboxylic acid, acmc-1bwzv, 3-isothiazolecarboxylicacid,4,5-dichloro, 4,5-dichloro-3-isothiazolecarboxylic acid
Formula Weight 198.03
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloroisothiazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.