missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichlorophthalic anhydride, 98%

Product Code. 10227793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227793 25 g Glass bottle 25g
10686824 1 g Glass bottle 1g
10208223 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10227793 Supplier Thermo Scientific Acros Supplier No. 387120250
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 942-06-3 | C8H2Cl2O3 | 217.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-06-3
Molecular Formula C8H2Cl2O3
Molecular Weight (g/mol) 217.00
MDL Number MFCD00075034
InChI Key ULSOWUBMELTORB-UHFFFAOYSA-N
Synonym 4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride
PubChem CID 70334
IUPAC Name 5,6-dichloro-2-benzofuran-1,3-dione
SMILES ClC1=CC2=C(C=C1Cl)C(=O)OC2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 185°C to 187°C
Color Blue to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 217.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 5-Dichlorophthalic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dichlorophthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.