missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyanoimidazole, 98+%

Product Code. 11348675
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348675 5 g 5g
11358675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348675 Supplier Thermo Scientific Alfa Aesar Supplier No. L12215.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1122-28-7 | C5H2N4 | 118.10 g/mol

4,5-Dicyanoimidazole is used in activation of nucleoside phosphoramidites during solid-phase oligonucleotide synthesis, and synthesis of nucleoside phosphoramidites, synthesis of novel nucleosides. It is often used as an alternative to tetrazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dicyanoimidazole is used in activation of nucleoside phosphoramidites during solid-phase oligonucleotide synthesis, and synthesis of nucleoside phosphoramidites, synthesis of novel nucleosides. It is often used as an alternative to tetrazole.

Solubility
Soluble in Acetonitrile: 1.8g/10ml, DMSO, Methanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-28-7
Molecular Formula C5H2N4
Molecular Weight (g/mol) 118.10
MDL Number MFCD00005194
InChI Key XGDRLCRGKUCBQL-UHFFFAOYSA-N
PubChem CID 70729
IUPAC Name 1H-imidazole-4,5-dicarbonitrile
SMILES N#CC1=C(N=CN1)C#N

Specifications

Melting Point 173°C to 177°C
Odor Odorless
Quantity 5 g
Beilstein 118208
Synonym 4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole
Solubility Information Soluble in Acetonitrile: 1.8g/10ml,DMSO,Methanol.
Formula Weight 118.1
Percent Purity ≥98%
Chemical Name or Material 4,5-Dicyanoimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.