missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitroaniline, TRC

Product Code. 30374747
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374747 1 g
30375434 500 mg
30388515 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374747 Supplier Toronto Research Chemicals Supplier No. TRCD4606451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7595-31-5
Molecular Formula C8H10N2O4
Molecular Weight (g/mol) 198.18
Synonym 4,5-Dimethoxy-2-nitro-benzenamine, 4,5-Dimethoxy-2-nitro-aniline, 4-Amino-5-nitroveratrole
SMILES COc1cc(N)c(cc1OC)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H10N2O4/c1-13-7-3-5(9)6(10(11)12)4-8(7)14-2/h3-4H,9H2,1-2H3
Formula Weight 198.0641
Chemical Name or Material 4,5-Dimethoxy-2-nitroaniline
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.