missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-o-phenylenediamine, 98%

Product Code. 11905271 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11905271

Brand: Thermo Scientific Acros 116610250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3171-45-7 | C8H12N2 | 136.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3171-45-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007729
InChI Key XSZYBMMYQCYIPC-UHFFFAOYSA-N
Synonym 4,5-dimethyl-1,2-phenylenediamine, 4,5-diamino-o-xylene, 4,5-dimethyl-o-phenylenediamine, 1,2-benzenediamine, 4,5-dimethyl, 1,2-diamino-4,5-dimethylbenzene, 4,5-dimethyl-1,2-benzenediamine, 4,5-dimethyl-benzene-1,2-diamine, 4,5-dimethyl-ortho-phenylenediamine, 4,5-dimethylphenylene-1,2-diamine, 4,5-dimethyl-o-phenylenediamine monohydrate
PubChem CID 76635
IUPAC Name 4,5-dimethylbenzene-1,2-diamine
SMILES CC1=CC(N)=C(N)C=C1C

Specifications

Melting Point 127.2°C to 128.3°C
CAS Min % 97.5
Color Brown to White
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Mass, Crystals or Chunks
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)2C6H2(NH2)2
Beilstein 13,179
Solubility Information Solubility in water: slightly soluble. Other solubilities: good soluble in acidified water,soluble in alcohols and polar solvents
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 4, 5-Dimethyl-o-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dimethyl-o-phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.