missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethylthiazole, 98%

Product Code. 11499692
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499692 5 g 5g
11409702 25 g 25g
11419702 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499692 Supplier Thermo Scientific Alfa Aesar Supplier No. A10785.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3581-91-7 | C5H7NS | 113.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3581-91-7
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005336
InChI Key UWSONZCNXUSTKW-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazole, thiazole, 4,5-dimethyl, 4,5-dimethyl thiazole, 4,5-dimethyl-thiazole, unii-u3rp5i088g, fema no. 3274, pubchem9938, acmc-209ijb, dimethylthiazole, 4,5, 4-27-00-00986 beilstein handbook reference
PubChem CID 62510
IUPAC Name 4,5-dimethyl-1,3-thiazole
SMILES CC1=C(SC=N1)C

Specifications

Density 1.07
Boiling Point 157°C to 158°C
Flash Point 51°C (123°F)
Refractive Index 1.521
Quantity 5 g
UN Number UN1993
Beilstein 105694
Formula Weight 113.18
Percent Purity 98%
Chemical Name or Material 4,5-Dimethylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.