missing translation for 'onlineSavingsMsg'
Learn More

4,5-Epoxy-13-ethyl-17-hydroxy-18,19-dinor-17Alpha-pregn-20-yn-3-one, TRC

Product Code. 30378787
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378787 250 mg
30369693 25 mg
30368800 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378787 Supplier Toronto Research Chemicals Supplier No. TRCE891940250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25073-30-7
Molecular Formula C21H28O3
Molecular Weight (g/mol) 328.45
Synonym Levonorgestrel Impurity 7 (Mixture of Diastereomers)
SMILES CC[C@]12CC[C@H]3[C@@H](CCC45OC4C(=O)CC[C@H]35)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H28O3/c1-3-19-10-7-14-13(15(19)9-11-20(19,23)4-2)8-12-21-16(14)5-6-17(22)18(21)24-21/h2,13-16,18,23H,3,5-12H2,1H3/t13-,14+,15+,16-,18?,19+,20+,21?/m1/s1
Formula Weight 328.2038
Chemical Name or Material 4,5-Epoxy-13-ethyl-17-hydroxy-18,19-dinor-17alpha-pregn-20-yn-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.