missing translation for 'onlineSavingsMsg'
Learn More

4,5-Imidazoledicarboxylic acid, 97%

Product Code. 11473378
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473378 100 g 100g
11453378 5 g 5g
11463378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473378 Supplier Thermo Scientific Alfa Aesar Supplier No. A17783.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 570-22-9 | C5H4N2O4 | 156.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-22-9
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00005200
InChI Key ZEVWQFWTGHFIDH-UHFFFAOYSA-N
Synonym 4,5-imidazoledicarboxylic acid, imidazole-4,5-dicarboxylic acid, glyoxalinedicarboxylic acid, 4,5-dicarboxyimidazole, 4,5-imidazoledicarboxylate, glycoxalinedicarboxylic acid, alpha-beta-imidazolecarboxylic acid, alpha,beta-imidazoledicarboxylic acid, 4.5-imidazoledicarboxylic acid, 4,5-imidazole dicarboxylic acid
PubChem CID 68442
IUPAC Name 1H-imidazole-4,5-dicarboxylic acid
SMILES OC(=O)C1=C(N=CN1)C(O)=O

Specifications

Melting Point ∼283°C (decomposition)
Density 1.749
Quantity 100 g
Beilstein 147774
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 4,5-Imidazoledicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.