missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10094322
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10094322 250 mg Amber Glass Bottle 250mg
10197842 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10094322 Supplier Thermo Scientific Maybridge Supplier No. CC34801CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 95124-68-8
Molecular Formula C10H8N2O3
Molecular Weight (g/mol) 204.185
MDL Number MFCD06797475
InChI Key DTKAUYQBZSOHRJ-UHFFFAOYSA-N
Synonym 4-5-methyl-1,2,4-oxadiazol-3-yl benzoic acid, 3-4-carboxyphenyl-5-methyl-1,2,4-oxadiazole, 4-5-methyl-1,2,4 oxadiazol-3-yl-benzoic acid, benzoic acid, 4-5-methyl-1,2,4-oxadiazol-3-yl, acmc-209xiw, 4-5-methyl-1,2,4-oxadiazol-3-yl benzoicacid
PubChem CID 7127808
IUPAC Name 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoic acid
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.18
Percent Purity 97%
Chemical Name or Material 4-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.