missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15409995
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409995 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15409995 Supplier Thermo Scientific Alfa Aesar Supplier No. H52658.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 913835-70-8
Melting Point 188°C to 190°C
Molecular Formula C9H9BN2O3
MDL Number MFCD08436062
Quantity 250 mg
Synonym 4-5-methyl-1,3,4-oxadiazol-2-yl phenylboronic acid, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl boronic acid, 2-4-boronophenyl-5-methyl-1,3,4-oxadiazole, boronicacid, b-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl, 4-5-methyl-1,3,4-oxadiazol-2-yl benzeneboronic acid, acmc-209r9v, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl boronicacid
InChI Key SPSKFVHUUJWUPB-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=NN=C(O2)C)(O)O
IUPAC Name [4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]boronic acid
Molecular Weight (g/mol) 203.992
PubChem CID 44119343
Formula Weight 203.99
Percent Purity 98%
Chemical Name or Material 4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.