missing translation for 'onlineSavingsMsg'
Learn More

4,(5)-Methyl Mercapto Benzimidazole Zinc Salt, TRC

Product Code. 30484160
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484160 10 g 10g
30474656 5 g 5g
30483781 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484160 Supplier Toronto Research Chemicals Supplier No. TRCM23787010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61617-00-3
Molecular Formula C16H14N4S2Zn
Molecular Weight (g/mol) 391.82
Synonym Antioxidant ZMMBI, Nocrac MMBZ, Rhenogran ZMMBI 50, Vanox ZMTI, Vulkanox ZMB 2, Vulkanox ZMBI, ZMMBI, ZMTI, Zinc 2-Mercaptotoluimidazole, Zinc Methylmercaptobenzimidazole
IUPAC Name bis[(4-methyl-1H-benzimidazol-2-yl)sulfanyl]zinc
SMILES Cc1cccc2[nH]c(S[Zn]Sc3nc4c(C)cccc4[nH]3)nc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/2C8H8N2S.Zn/c2*1-5-3-2-4-6-7(5)10-8(11)9-6;/h2*2-4H,1H3,(H2,9,10,11);/q;;+2/p-2
Formula Weight 389.9951
Chemical Name or Material 4,(5)-Methyl Mercapto Benzimidazole Zinc Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.