missing translation for 'onlineSavingsMsg'
Learn More

4',5'-Methylenedioxy-2'-nitroacetophenone, 96%

Product Code. 11332657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332657 1 g 1g
11342657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332657 Supplier Thermo Scientific Alfa Aesar Supplier No. L11841.03
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 56136-84-6 | C9H7NO5 | 209.157 g/mol

4',5'-Methylenedioxy-2'-nitroacetophenone is used as an intermediate in organic syntheses and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5′-Methylenedioxy-2′-nitroacetophenone is used as an intermediate in organic syntheses and in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56136-84-6
Molecular Formula C9H7NO5
Molecular Weight (g/mol) 209.157
MDL Number MFCD00053045
InChI Key BQONDGIXVHVIIR-UHFFFAOYSA-N
Synonym 1-6-nitro-1,3-benzodioxol-5-yl ethanone, 4',5'-methylenedioxy-2'-nitroacetophenone, 4,5-methylenedioxy-2-nitroacetophenone, 1-6-nitrobenzo d 1,3 dioxol-5-yl ethanone, 1-6-nitro-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethan-1-one, 1-6-nitro-2h-1,3-benzodioxol-5-yl ethanone, 5-acetyl-6-nitro-1,3-benzodioxole, 4.5-methylenedioxy-2-nitroacetophenone, 5-acetyl-6-nitro-2h-benzo d 1,3-dioxolene
PubChem CID 92022
IUPAC Name 1-(6-nitro-1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1[N+](=O)[O-])OCO2

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 199756
Solubility Information Insoluble in water.
Formula Weight 209.16
Percent Purity 98%
Chemical Name or Material 4',5'-Methylenedioxy-2'-nitroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.