missing translation for 'onlineSavingsMsg'
Learn More

{4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine, 97%, Thermo Scientific™

Product Code. 10612675
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10612675 1 g Amber Glass Bottle 1g
10608534 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10612675 Supplier Thermo Scientific Maybridge Supplier No. CC53013DA
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-74-7
Molecular Formula C13H11F3N2
Molecular Weight (g/mol) 252.24
MDL Number MFCD09817478
InChI Key XQAJYHFQYZEWSE-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl benzylamine, 4-5-trifluoromethyl pyrid-2-yl phenyl methylamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl pyridin-2-yl phenyl methylamine, 1-4-5-trifluoromethyl pyridin-2-yl phenyl methanamine, 4-5-trifluoromethyl-2-pyridyl phenyl methylamine, 2-4-aminomethyl phenyl-5-trifluoromethyl pyridine
PubChem CID 24229534
IUPAC Name [4-[5-(trifluoromethyl)pyridin-2-yl]phenyl]methanamine
SMILES C1=CC(=CC=C1CN)C2=NC=C(C=C2)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 252.24
Percent Purity 97%
Chemical Name or Material {4-[5-(Trifluoromethyl)pyrid-2-yl]phenyl}methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.