missing translation for 'onlineSavingsMsg'
Learn More

4',6-Diamidino-2-phenylindole dihydrochloride, >98%, MP Biomedicals™

Product Code. 11421021 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421021 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421021 Supplier MP Biomedicals Supplier No. 0215757410
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
IUPAC Name dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

Color Yellow to Orange
Assay Percent Range >98%
Quantity 10 mg
Formula Weight 350.2
Physical Form Crystalline Powder
Chemical Name or Material 4', 6-Diamidino-2-phenylindole dihydrochloride
compliance-icons
Product Identifier
  • 4',6-Diamidino-2-phenylindole Dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: DAPI dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.