missing translation for 'onlineSavingsMsg'
Learn More

4',6'-Diamidino-2-phenylindole dihydrochloride, 98%

Product Code. 11916621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916621 10 mg Glass bottle 10mg
11926621 50 mg Glass bottle 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916621 Supplier Thermo Scientific Acros Supplier No. 202710100
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28718-90-3 | C16H17Cl2N5 | 350.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information (1% in water) Clear
IUPAC Name 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide;dihydrochloride
Formula Weight 350.25
Percent Purity 98%
Physical Form Solution
Chemical Name or Material 4′,6′-Diamidino-2-phenylindole dihydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.