missing translation for 'onlineSavingsMsg'
Learn More

4’,6-Diamidino-2-phenylindole Dihydrochloride, TRC

Product Code. 30134919
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134919 25 mg 25mg
30012370 100 mg 100mg
17962585 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134919 Supplier Toronto Research Chemicals Supplier No. TRCD41605025MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16 H15 N5 . 2 Cl H
Molecular Weight (g/mol) 350.25
Synonym DAPI
IUPAC Name 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide;dihydrochloride
SMILES Cl.Cl.NC(=N)c1ccc(cc1)c2cc3ccc(cc3[nH]2)C(=N)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H15N5.2ClH/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;;/h1-8,21H,(H3,17,18)(H3,19,20);2*1H
Formula Weight 349.0861
Chemical Name or Material 4',6-Diamidino-2-phenylindole Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.