missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diamino-2-hydroxy-1,3,5-triazine, tech. 90%

Product Code. 11317866
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317866 1 g 1g
11327866 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317866 Supplier Thermo Scientific Alfa Aesar Supplier No. L13064.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 645-92-1 | C3H5N5O | 127.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-92-1
Molecular Formula C3H5N5O
Molecular Weight (g/mol) 127.107
MDL Number MFCD00023186
InChI Key MASBWURJQFFLOO-UHFFFAOYSA-N
Synonym ammeline, 4,6-diamino-1,3,5-triazin-2-ol, ammelin, 1,3,5-triazin-2 1h-one, 4,6-diamino, 2,4-diamino-6-hydroxy-1,3,5-triazine, cyanurodiamide, 2,4-diamino-1,3,5-triazin-6-one, 4,6-diamino-1,3,5-triazin-2 1h-one, unii-lc8g9556q0, s-triazin-2-ol, 4,6-diamino
PubChem CID 12583
ChEBI CHEBI:28646
IUPAC Name 2,6-diamino-1H-1,3,5-triazin-4-one
SMILES C1(=NC(=O)N=C(N1)N)N

Specifications

Quantity 1 g
Beilstein 5570
Formula Weight 127.11
Percent Purity 90%
Grade Technical
Chemical Name or Material 4,6-Diamino-2-hydroxy-1,3,5-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.