missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diamino-2-thiopyrimidine, TRC

Product Code. 30180126
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180126 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180126 Supplier Toronto Research Chemicals Supplier No. TRCD41686050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1004-39-3
Molecular Formula C4 H6 N4 S
Quantity 50 g
Synonym 2(1H)-Pyrimidinethione, 4,6-diamino-, 2-Pyrimidinethiol, 4,6-diamino- (6CI), 4,6-Diamino-2(1H)-pyrimidinethione, 2-Mercapto-4,6-diaminopyrimidine, 2-Thio-6-aminocytosine, 4,6-Diamino-1,2-dihydro-2-thioxopyrimidine, 4,6-Diamino-1H-pyrimidine-2-thione, 4,6-Diamino-2-mercaptopyrimidine, 4,6-Diamino-2-pyrimidinethiol, 4,6-Diamino-2-thiopyrimidine, 4,6-Diamino-2-thiouracil, 4,6-Diaminopyrimidine-2(1H)-thione, 4,6-Diaminopyrimidine-2-thione, NSC 1586, NSC 680831
InChI Formula InChI=1S/C4H6N4S/c5-2-1-3(6)8-4(9)7-2/h1H,(H5,5,6,7,8,9)
SMILES NC1=CC(=NC(=S)N1)N
IUPAC Name 4,6-diamino-1H-pyrimidine-2-thione
Molecular Weight (g/mol) 142.18
Formula Weight 142.0313
Chemical Name or Material 4,6-Diamino-2-thiopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.