missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-methylpyrimidine, 97%, Thermo Scientific™

Product Code. 10342210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10342210 5 g Glass bottle 5g
10872071 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10342210 Supplier Thermo Scientific Acros Supplier No. 434260050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1780-26-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.01
MDL Number MFCD00090472
InChI Key FIMUTBLUWQGTIJ-UHFFFAOYSA-N
Synonym 2-methyl-4,6-dichloropyrimidine, 4,6-dichloro-2-methyl-pyrimidine, 2-mdcp, pyrimidine, 4,6-dichloro-2-methyl, 4,6-dichloro-2-methylpyrimidiine, 4,6-dichlor-2-methylpyrimidin, 4,6-dichloro-2-methyl pyrimidine, pubchem7075, acmc-1bpg6, ksc490s3h
PubChem CID 234997
IUPAC Name 4,6-dichloro-2-methylpyrimidine
SMILES CC1=NC(=CC(=N1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 45.7°C to 47.3°C
Color White
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 163.01
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4, 6-Dichloro-2-methylpyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dichloro-2-methylpyrimidine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.