missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-fluoropyrimidine, 98%

Product Code. 15478338
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478338 1 g 1g
15488338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478338 Supplier Thermo Scientific Alfa Aesar Supplier No. H61770.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 213265-83-9 | C4HCl2FN2 | 166.96 g/mol

4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213265-83-9
Molecular Formula C4HCl2FN2
Molecular Weight (g/mol) 166.96
MDL Number MFCD08056331
InChI Key DGMIGAHDDPJOPN-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro
PubChem CID 11286675
IUPAC Name 4,6-dichloro-5-fluoropyrimidine
SMILES FC1=C(Cl)N=CN=C1Cl

Specifications

Boiling Point 50°C to 60°C (14 mmHg)
Flash Point 91°C (196°F)
Quantity 1 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 166.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4,6-Dichloro-5-fluoropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.