missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine-5-carboxylic acid, 95%

Product Code. 15490969
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490969 1 g 1g
15410979 250 mg 250mg
15400979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490969 Supplier Thermo Scientific Alfa Aesar Supplier No. H63976.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87600-98-4 | C5H2Cl2N2O2 | 192.983 g/mol

4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87600-98-4
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD09909611
InChI Key KGBLYFGHJNGQBK-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro
PubChem CID 24820487
IUPAC Name 4,6-dichloropyrimidine-5-carboxylic acid
SMILES C1=NC(=C(C(=N1)Cl)C(=O)O)Cl

Specifications

Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 192.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,6-Dichloropyrimidine-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.