missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-2-mercaptopyrimidine, 98%

Product Code. 11400573
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400573 25 g 25g
11410573 100 g 100g
11420573 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400573 Supplier Thermo Scientific Alfa Aesar Supplier No. A12681.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol

4,6-Dihydroxy-2-mercaptopyrimidine is used as a reagent in assaying malondialdehyde (theTBARS assay of lipid peroxidation). It is used to detect lipid oxidation, quantitating sialic acids and lipopolysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-17-6
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
MDL Number MFCD00006674
InChI Key RVBUGGBMJDPOST-UHFFFAOYSA-N
Synonym 2-thiobarbituric acid, thiobarbituric acid, bathyran, austranal, 4,6-dihydroxy-2-thiopyrimidine, 2-thioxodihydropyrimidine-4,6 1h,5h-dione, 2-thio-4,6-dioxypyrimidine, usaf ek-660, 4,6 1h,5h-pyrimidinedione, dihydro-2-thioxo, 2-mercaptobarbituric acid
PubChem CID 2723628
ChEBI CHEBI:33202
IUPAC Name 2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES O=C1CC(=O)NC(=S)N1

Specifications

Melting Point >300°C
pH 1.8
Quantity 25 g
Beilstein 120663
Formula Weight 144.15
Percent Purity 98%
Chemical Name or Material 4,6-Dihydroxy-2-mercaptopyrimidine
compliance-icons
Product Identifier
  • 4,6-Dihydroxy-2-mercaptopyrimidin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4,6-Dihydroxy-2-mercaptopyrimidin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.