missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dihydroxy-5-nitropyrimidine, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | 4,6-Dihydroxy-5-nitropyrimidine |
| CAS | 2164-83-2 |
| Molecular Formula | C4 H3 N3 O4 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C4H3N3O4/c8-3-2(7(10)11)4(9)6-1-5-3/h1H,(H2,5,6,8,9) |
| SMILES | Oc1ncnc(O)c1[N+](=O)[O-] |
| IUPAC Name | 5-nitropyrimidine-4,6-diol |
| Molecular Weight (g/mol) | 157.08 |
| Formula Weight | 157.0124 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction