missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-pyrone, 98%

Product Code. 10157713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157713 1 g Glass bottle 1g
10115940 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157713 Supplier Thermo Scientific Acros Supplier No. 295350010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 675-09-2 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-09-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00075555
Synonym 4,6-dimethyl-2-pyrone, 4,6-dimethyl-2h-pyran-2-one, mesitene lactone, 4,6-dimethylcoumalin, 2h-pyran-2-one, 4,6-dimethyl, 4,6-dimethyl-alpha-pyrone, 4,6-dimethyl-pyran-2-one, 2,4-dimethyl-alpha-pyrone, 2,4-dimethyl-.alpha.-pyrone, 4,6-dimethyl-.alpha.-pyrone

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 50°C
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 124.14
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4, 6-Dimethyl-2-pyrone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.