missing translation for 'onlineSavingsMsg'
Learn More

4,6-dimethyl-3-nitropyridin-2(1{H})-one, TRC

Product Code. 30408525
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408525 50 mg
30391981 100 mg
30406506 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408525 Supplier Toronto Research Chemicals Supplier No. TRCH66959550MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22934-13-0
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
Synonym 2-Pyridinol, 4,6-dimethyl-3-nitro- (7CI,8CI), 4,6-Dimethyl-3-nitro-2(1H)-pyridinone
IUPAC Name 4,6-dimethyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC(=C(C(=O)N1)[N+](=O)[O-])C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H8N2O3/c1-4-3-5(2)8-7(10)6(4)9(11)12/h3H,1-2H3,(H,8,10)
Formula Weight 168.0535
Chemical Name or Material 4,6-dimethyl-3-nitropyridin-2(1(H))-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.